4-Isobutoxy-3-(trifluoromethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

76801-500MG 76801-1G 76801-5G
77335-594EA 359.47 USD
77335-594 77358-454 77382-338
4-Isobutoxy-3-(trifluoromethyl)benzaldehyde ≥97%
4-Isobutoxy-3-(trifluoromethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₂H₁₃F₃O₂
MW: 246.226 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19384245
CAS Number: 1487788-21-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR