1-Bromo-3-chloro-2-(methylsulfonyl)-4-propoxybenzene ≥95%
Supplier: AOBChem USA
|
Warning
|
76749-500MG
76749-1G
76749-5G
76749-25G
77334-256EA
204.97
USD
77334-256
77358-450
77370-266
77391-058
1-Bromo-3-chloro-2-(methylsulfonyl)-4-propoxybenzene ≥95%
1-Bromo-3-chloro-2-(methylsulfonyl)-4-propoxybenzene
Precaución: for Research Use Only
Formula:
C₁₀H₁₂BrClO₃S MW: 327.622 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34766699 CAS Number: 2901099-41-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...