5-Bromo-2-fluoro-1-iodo-3-(methylsulfonyl)benzene ≥97%

Supplier: AOBChem USA

Warning

76741-500MG 76741-1G 76741-5G 76741-25G
77334-252EA 281.19 USD
77334-252 77358-446 77370-260 77391-054
5-Bromo-2-fluoro-1-iodo-3-(methylsulfonyl)benzene ≥97%
5-Bromo-2-fluoro-1-iodo-3-(methylsulfonyl)benzene

Precaución: for Research Use Only

Formula: C₇H₅BrFIO₂S
MW: 378.985 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34766695
CAS Number: 2901083-38-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR