2,4-Dibromo-6-methoxyphenol ≥97%

Supplier: AOBChem USA

Warning

76727-500MG 76727-1G 76727-5G
77337-012EA 404.79 USD
77337-012 77358-442 77382-330
2,4-Dibromo-6-methoxyphenol ≥97%
2,4-Dibromo-6-methoxyphenol

Precaución: for Research Use Only

Formula: C₇H₆Br₂O₂
MW: 281.929 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00029691
CAS Number: 53948-36-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR