(6-(Benzyloxy)-2,3-difluorophenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

76681-500MG 76681-1G 76681-5G 76681-25G
77334-248EA 162.74 USD
77334-248 77346-100 77370-256 77391-050
(6-(Benzyloxy)-2,3-difluorophenyl)(methyl)sulfane ≥97%
(6-(Benzyloxy)-2,3-difluorophenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₁₄H₁₂F₂OS
Storage Temperature: Ambient
MDL Number: MFCD34766675
CAS Number: 2901065-51-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR