1-(Benzyloxy)-3,4-difluoro-2-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

76680-500MG 76680-1G 76680-5G 76680-25G
77334-246EA 219.39 USD
77334-246 77358-440 77370-254 77391-048
1-(Benzyloxy)-3,4-difluoro-2-iodobenzene ≥97%
1-(Benzyloxy)-3,4-difluoro-2-iodobenzene

Precaución: for Research Use Only

Formula: C₁₃H₉F₂IO
Storage Temperature: Ambient
MDL Number: MFCD34766674
CAS Number: 2901107-10-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR