1-(Benzyloxy)-2-bromo-3,4-difluorobenzene ≥97%

Supplier: AOBChem USA

Warning

76679-500MG 76679-1G 76679-5G 76679-25G
77334-244EA 194.67 USD
77334-244 77358-438 77370-252 77391-046
1-(Benzyloxy)-2-bromo-3,4-difluorobenzene ≥97%
1-(Benzyloxy)-2-bromo-3,4-difluorobenzene

Precaución: for Research Use Only

Formula: C₁₃H₉BrF₂O
MW: 299.111 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34766673
CAS Number: 1376335-06-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR