3-Methyl-5-(methylthio)benzonitrile ≥97%

Supplier: AOBChem USA

Warning

76648-1G 76648-5G 76648-25G
77341-850EA 105.06 USD
77341-850 77370-250 77382-328
3-Methyl-5-(methylthio)benzonitrile ≥97%
3-Methyl-5-(methylthio)benzonitrile

Precaución: for Research Use Only

Formula: C₉H₉NS
Storage Temperature: Ambient
MDL Number: MFCD22489960
CAS Number: 1822671-99-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR