1-Bromo-2-(methoxymethyl)-4,5-dimethylbenzene ≥97%

Supplier: AOBChem USA

Warning

76543-500MG 76543-1G 76543-5G
77337-236EA 408.91 USD
77337-236 77358-434 77382-324
1-Bromo-2-(methoxymethyl)-4,5-dimethylbenzene ≥97%
1-Bromo-2-(methoxymethyl)-4,5-dimethylbenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₃BrO
MW: 229.1136 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34766592
CAS Number: 2901067-07-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR