2-Isobutoxy-4,5-dimethylbenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

76539-500MG 76539-1G 76539-5G
77339-942EA 685.98 USD
77339-942 77358-432 77382-322
2-Isobutoxy-4,5-dimethylbenzaldehyde ≥95%
2-Isobutoxy-4,5-dimethylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₁₈O₂
MW: 206.281 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34766588
CAS Number: 2901081-64-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR