2-Isobutoxy-4,5-dimethylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

76537-500MG 76537-1G 76537-5G
77337-550EA 420.24 USD
77337-550 77358-430 77382-320
2-Isobutoxy-4,5-dimethylbenzoic acid ≥97%
2-Isobutoxy-4,5-dimethylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₈O₃
MW: 222.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34766586
CAS Number: 2901099-25-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR