1-Fluoro-2-(methoxymethyl)-4-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

76525-1G 76525-5G 76525-25G
77341-848EA 105.06 USD
77341-848 77370-246 77382-316
1-Fluoro-2-(methoxymethyl)-4-methylbenzene ≥97%
1-Fluoro-2-(methoxymethyl)-4-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₁FO
MW: 154.181 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34766583
CAS Number: 2901099-95-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR