1-Bromo-2-chloro-3-(methoxymethyl)-5-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

76507-500MG 76507-1G 76507-5G 76507-25G
77334-240EA 162.74 USD
77334-240 77346-098 77370-244 77391-042
1-Bromo-2-chloro-3-(methoxymethyl)-5-methylbenzene ≥97%
1-Bromo-2-chloro-3-(methoxymethyl)-5-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₀BrClO
MW: 249.532 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34766565
CAS Number: 2901102-93-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR