2,4-Difluoro-5-methyl-3-(trimethylsilyl)benzaldehyde ≥97%
Supplier: AOBChem USA
|
Warning
|
76399-500MG
76399-1G
76399-5G
76399-25G
77334-232EA
122.57
USD
77334-232
77344-012
77370-238
77382-308
2,4-Difluoro-5-methyl-3-(trimethylsilyl)benzaldehyde ≥97%
2,4-Difluoro-5-methyl-3-(trimethylsilyl)benzaldehyde
Precaución: for Research Use Only
Formula:
C₁₁H₁₄F₂OSi MW: 228.3105 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34766505 CAS Number: 348081-00-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...