4-Fluoro-3-methyl-5-(methylthio)phenol ≥97%

Supplier: AOBChem USA

Warning

76369-500MG 76369-1G 76369-5G 76369-25G
77334-230EA 260.59 USD
77334-230 77358-418 77370-236 77391-036
4-Fluoro-3-methyl-5-(methylthio)phenol ≥97%
4-Fluoro-3-methyl-5-(methylthio)phenol

Precaución: for Research Use Only

Formula: C₈H₉FOS
MW: 172.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34766486
CAS Number: 2710987-61-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR