2-Fluoro-5-hydroxy-3-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

76362-500MG 76362-1G 76362-5G 76362-25G
77334-228EA 275.01 USD
77334-228 77358-416 77370-234 77391-034
2-Fluoro-5-hydroxy-3-methylbenzoic acid ≥97%
2-Fluoro-5-hydroxy-3-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₇FO₃
Storage Temperature: Ambient
MDL Number: MFCD34766480
CAS Number: 1894825-45-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR