2-Fluoro-1-iodo-5-isopropoxy-3-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

76350-500MG 76350-1G 76350-5G 76350-25G
77334-226EA 280.16 USD
77334-226 77358-414 77370-232 77391-032
2-Fluoro-1-iodo-5-isopropoxy-3-methylbenzene ≥97%
2-Fluoro-1-iodo-5-isopropoxy-3-methylbenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₂FIO
MW: 294.105 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34766472
CAS Number: 2918896-38-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR