2-Fluoro-5-isopropoxy-3-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

76348-500MG 76348-1G 76348-5G 76348-25G
77334-224EA 252.35 USD
77334-224 77358-412 77370-230 77391-030
2-Fluoro-5-isopropoxy-3-methylbenzaldehyde ≥97%
2-Fluoro-5-isopropoxy-3-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₃FO₂
Storage Temperature: Ambient
MDL Number: MFCD34766470
CAS Number: 2901098-45-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR