2-Fluoro-5-isopropoxy-3-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

76346-500MG 76346-1G 76346-5G 76346-25G
77334-222EA 146.26 USD
77334-222 77345-144 77370-228 77391-028
2-Fluoro-5-isopropoxy-3-methylbenzoic acid ≥97%
2-Fluoro-5-isopropoxy-3-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₃FO₃
Storage Temperature: Ambient
MDL Number: MFCD34766468
CAS Number: 2055398-41-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR