5-Chloro-6-ethoxy-[1,1'-biphenyl]-3-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

76302-500MG 76302-1G 76302-5G 76302-25G
77334-218EA 273.98 USD
77334-218 77358-408 77370-224 77391-024
5-Chloro-6-ethoxy-[1,1'-biphenyl]-3-carboxylic acid ≥97%
5-Chloro-6-ethoxy-[1,1'-biphenyl]-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₃ClO₃
MW: 276.715 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34766425
CAS Number: 2901102-76-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR