2-(Benzyloxy)-5-bromo-3-chloro-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

76299-1G 76299-5G 76299-25G
77341-846EA 105.06 USD
77341-846 77370-222 77382-304
2-(Benzyloxy)-5-bromo-3-chloro-1,1'-biphenyl ≥97%
2-(Benzyloxy)-5-bromo-3-chloro-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₉H₁₄BrClO
MW: 373.671 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34766422
CAS Number: 2901105-41-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR