4-Isobutoxy-2,5-dimethylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

76240-500MG 76240-1G 76240-5G 76240-25G
77334-212EA 182.31 USD
77334-212 77347-042 77370-218 77391-022
4-Isobutoxy-2,5-dimethylbenzaldehyde ≥97%
4-Isobutoxy-2,5-dimethylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₁₈O₂
MW: 206.281 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34766368
CAS Number: 1538202-28-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR