4-Isopropoxy-2,5-dimethylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

76224-500MG 76224-1G 76224-5G
77337-234EA 408.91 USD
77337-234 77358-398 77382-296
4-Isopropoxy-2,5-dimethylbenzoic acid ≥97%
4-Isopropoxy-2,5-dimethylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₃
MW: 208.254 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14537796
CAS Number: 1216114-70-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR