(4-Isopropoxy-2,5-dimethylphenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

76222-500MG 76222-1G 76222-5G 76222-25G
77334-204EA 157.59 USD
77334-204 77345-682 77370-210 77391-016
(4-Isopropoxy-2,5-dimethylphenyl)boronic acid ≥97%
(4-Isopropoxy-2,5-dimethylphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₇BO₃
Storage Temperature: Ambient
MDL Number: MFCD34766354
CAS Number: 1644245-00-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR