1-(Benzyloxy)-4-iodo-2,5-dimethylbenzene ≥97%

Supplier: AOBChem USA

Warning

76218-500MG 76218-1G 76218-5G
77336-316EA 386.25 USD
77336-316 77358-396 77382-294
1-(Benzyloxy)-4-iodo-2,5-dimethylbenzene ≥97%
1-(Benzyloxy)-4-iodo-2,5-dimethylbenzene

Precaución: for Research Use Only

Formula: C₁₅H₁₅IO
MW: 338.183 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34766350
CAS Number: 2901083-23-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR