4-Acetyl-2,6-difluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

76203-500MG 76203-1G 76203-5G 76203-25G
77334-200EA 108.15 USD
77334-200 77343-208 77370-204 77382-288
4-Acetyl-2,6-difluorobenzoic acid ≥97%
4-Acetyl-2,6-difluorobenzoic acid

Precaución: for Research Use Only

Formula: C₉H₆F₂O₃
Storage Temperature: Ambient
MDL Number: MFCD26687993
CAS Number: 1518217-36-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR