4-Chloro-1-ethyl-2-fluorobenzene ≥97%

Supplier: AOBChem USA

Warning

76176-500MG 76176-1G 76176-5G 76176-25G
77334-196EA 135.96 USD
77334-196 77344-520 77370-200 77391-012
4-Chloro-1-ethyl-2-fluorobenzene ≥97%
4-Chloro-1-ethyl-2-fluorobenzene

Precaución: for Research Use Only

Formula: C₈H₈ClF
Storage Temperature: Ambient
MDL Number: MFCD29091077
CAS Number: 1369880-08-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR