4-Chloro-1-ethoxy-2-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

76133-500MG 76133-1G 76133-5G 76133-25G
77334-194EA 192.61 USD
77334-194 77358-390 77370-198 77391-010
4-Chloro-1-ethoxy-2-methylbenzene ≥95%
4-Chloro-1-ethoxy-2-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₁ClO
MW: 170.636 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06252403
CAS Number: 35338-32-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR