5-(Benzyloxy)-4-fluoro-2-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

76084-500MG 76084-1G 76084-5G
77337-662EA 434.66 USD
77337-662 77358-384 77382-282
5-(Benzyloxy)-4-fluoro-2-methylbenzaldehyde ≥97%
5-(Benzyloxy)-4-fluoro-2-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₅H₁₃FO₂
Storage Temperature: Ambient
MDL Number: MFCD34766256
CAS Number: 2901082-59-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR