5-(Benzyloxy)-4-fluoro-2-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

76082-500MG 76082-1G 76082-5G
77340-106EA 757.05 USD
77340-106 77358-382 77382-280
5-(Benzyloxy)-4-fluoro-2-methylbenzoic acid ≥97%
5-(Benzyloxy)-4-fluoro-2-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₃FO₃
MW: 260.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31579465
CAS Number: 2270905-58-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR