4-Fluoro-5-isopropoxy-2-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

76073-500MG 76073-1G 76073-5G
77337-928EA 451.14 USD
77337-928 77358-378 77382-276
4-Fluoro-5-isopropoxy-2-methylbenzoic acid ≥97%
4-Fluoro-5-isopropoxy-2-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₃FO₃
MW: 212.218 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34766246
CAS Number: 2901095-48-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR