5-Ethoxy-4-fluoro-2-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

76064-500MG 76064-1G 76064-5G
77335-456EA 353.29 USD
77335-456 77358-374 77382-272
5-Ethoxy-4-fluoro-2-methylbenzoic acid ≥97%
5-Ethoxy-4-fluoro-2-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₃
Storage Temperature: Ambient
MDL Number: MFCD34766237
CAS Number: 2092609-63-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR