1-Bromo-5-ethoxy-4-fluoro-2-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

76061-500MG 76061-1G 76061-5G 76061-25G
77334-186EA 202.91 USD
77334-186 77358-370 77370-192 77391-004
1-Bromo-5-ethoxy-4-fluoro-2-methylbenzene ≥97%
1-Bromo-5-ethoxy-4-fluoro-2-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₀BrFO
MW: 233.078 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34766234
CAS Number: 2383768-46-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR