(4-Fluoro-5-methoxy-2-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

76059-500MG 76059-1G 76059-5G 76059-25G
77334-184EA 172.01 USD
77334-184 77346-426 77370-190 77391-002
(4-Fluoro-5-methoxy-2-methylphenyl)(methyl)sulfane ≥97%
(4-Fluoro-5-methoxy-2-methylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₉H₁₁FOS
MW: 186.246 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34766233
CAS Number: 2706073-76-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR