2-(2,5-Dichloropyridin-4-yl)acetic acid ≥97%

Supplier: AOBChem USA

Warning

76042-500MG 76042-1G 76042-5G
77339-328EA 578.86 USD
77339-328 77358-364 77382-264
2-(2,5-Dichloropyridin-4-yl)acetic acid ≥97%
2-(2,5-Dichloropyridin-4-yl)acetic acid

Precaución: for Research Use Only

Formula: C₇H₅Cl₂NO₂
MW: 206.0261 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16609367
CAS Number: 1173917-27-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR