2-(5-Bromo-2-chloropyridin-4-yl)acetic acid ≥95%

Supplier: AOBChem USA

Warning

76036-500MG 76036-1G 76036-5G 76036-25G
77334-180EA 204.97 USD
77334-180 77358-362 77370-188 77390-998
2-(5-Bromo-2-chloropyridin-4-yl)acetic acid ≥95%
2-(5-Bromo-2-chloropyridin-4-yl)acetic acid

Precaución: for Research Use Only

Formula: C₇H₅BrClNO₂
MW: 250.4771 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16609333
CAS Number: 1227592-37-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR