2-(5-Bromo-2-methoxypyridin-4-yl)acetic acid ≥97%

Supplier: AOBChem USA

Warning

76030-500MG 76030-1G 76030-25G 76030-5G
77334-178EA 297.67 USD
77334-178 77358-360 77390-996 77382-262
2-(5-Bromo-2-methoxypyridin-4-yl)acetic acid ≥97%
2-(5-Bromo-2-methoxypyridin-4-yl)acetic acid

Precaución: for Research Use Only

Formula: C₈H₈BrNO₃
MW: 246.0580 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16609439
CAS Number: 1227600-34-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR