2-Methyl-5-(methylthio)benzonitrile ≥97%

Supplier: AOBChem USA

Warning

75985-500MG 75985-1G 75985-25G 75985-5G
77334-176EA 284.28 USD
77334-176 77358-358 77390-994 77382-260
2-Methyl-5-(methylthio)benzonitrile ≥97%
2-Methyl-5-(methylthio)benzonitrile

Precaución: for Research Use Only

Formula: C₉H₉NS
Storage Temperature: Ambient
MDL Number: MFCD22488966
CAS Number: 1421631-78-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR