5-Bromo-3-methoxybenzene-1,2-diol ≥95%

Supplier: AOBChem USA

Warning

75886-500MG 75886-1G 75886-5G 75886-25G
77334-170EA 202.91 USD
77334-170 77358-354 77370-180 77390-988
5-Bromo-3-methoxybenzene-1,2-diol ≥95%
5-Bromo-3-methoxybenzene-1,2-diol

Precaución: for Research Use Only

Formula: C₇H₇BrO₃
MW: 219.0326 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00016723
CAS Number: 38790-07-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR