4-Bromo-2-fluoro-5-methoxy-3-(trimethylsilyl)benzoic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
75774-500MG
75774-1G
75774-5G
75774-25G
77334-166EA
82.4
USD
77334-166
77342-168
77370-172
77382-250
4-Bromo-2-fluoro-5-methoxy-3-(trimethylsilyl)benzoic acid ≥97%
4-Bromo-2-fluoro-5-methoxy-3-(trimethylsilyl)benzoic acid
Precaución: for Research Use Only
Formula:
C₁₁H₁₄BrFO₃Si MW: 321.215 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34766148 CAS Number: 2901102-44-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...