2-Bromo-4-fluoro-1-methoxy-3-(methylsulfonyl)benzene ≥97%

Supplier: AOBChem USA

Warning

75770-500MG 75770-1G 75770-5G 75770-25G
77334-162EA 182.31 USD
77334-162 77346-898 77370-170 77390-984
2-Bromo-4-fluoro-1-methoxy-3-(methylsulfonyl)benzene ≥97%
2-Bromo-4-fluoro-1-methoxy-3-(methylsulfonyl)benzene

Precaución: for Research Use Only

Formula: C₈H₈BrFO₃S
Storage Temperature: Ambient
MDL Number: MFCD34766144
CAS Number: 2901086-46-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR