2-Amino-5-iodo-3-methylbenzenecarboxylic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Amino-5-iodo-3-methylbenzoic acid

75742-1G 75742-5G 75742-25G
77340-822EA 60.77 USD
77340-822 77370-168 77382-244
2-Amino-5-iodo-3-methylbenzenecarboxylic acid ≥97%
2-Amino-5-iodo-3-methylbenzenecarboxylic acid

Precaución: for Research Use Only

Formula: C₈H₈INO₂
MW: 277.06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09029816
CAS Number: 108857-24-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR