5-Chloro-2-hydroxy-3-methylbenzoic acid

Supplier: AOBChem USA

Warning

75738-500MG 75738-1G 75738-5G 75738-25G
77334-158EA 155.53 USD
77334-158 77345-582 77370-166 77390-982
5-Chloro-2-hydroxy-3-methylbenzoic acid
5-Chloro-2-hydroxy-3-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₇ClO₃
MW: 186.5923 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06203201
CAS Number: 4386-40-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR