5-Bromo-2-chloro-3-methylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

75735-500MG 75735-1G 75735-5G
77336-288EA 384.19 USD
77336-288 77358-346 77382-242
5-Bromo-2-chloro-3-methylbenzoic acid ≥95%
5-Bromo-2-chloro-3-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆BrClO₂
MW: 249.4890 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18379819
CAS Number: 1263376-33-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR