Methyl 5-bromo-3-(trifluoromethyl)benzo[b]thiophene-2-carboxylate ≥95%

Supplier: AOBChem USA

Warning

75711-500MG 75711-1G 75711-5G
77337-010EA 404.79 USD
77337-010 77358-342 77382-240
Methyl 5-bromo-3-(trifluoromethyl)benzo[b]thiophene-2-carboxylate ≥95%
Methyl 5-bromo-3-(trifluoromethyl)benzo[b]thiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₁H₆BrF₃O₂S
Storage Temperature: Ambient
MDL Number: MFCD29195452
CAS Number: 617706-20-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR