5-(3-Methoxy-2-propoxyphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

75704-500MG 75704-1G 75704-25G 75704-5G
77334-156EA 301.79 USD
77334-156 77358-340 77390-980 77382-238
5-(3-Methoxy-2-propoxyphenyl)thiazol-2-amine ≥97%
5-(3-Methoxy-2-propoxyphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₃H₁₆N₂O₂S
MW: 264.3433 g/mol
Storage Temperature: Ambient
CAS Number: 2901107-07-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR