4-Fluoro-3-methoxy-5-(trifluoromethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

75694-500MG 75694-1G 75694-5G
77339-922EA 676.71 USD
77339-922 77358-338 77382-236
4-Fluoro-3-methoxy-5-(trifluoromethyl)benzaldehyde ≥97%
4-Fluoro-3-methoxy-5-(trifluoromethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₉H₆F₄O₂
MW: 222.1363 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34766111
CAS Number: 2385261-61-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR