Methyl 5-bromo-2-fluoro-3-(trifluoromethyl)benzoate ≥97%

Supplier: AOBChem USA

Warning

75669-500MG 75669-1G 75669-5G 75669-25G
77334-154EA 86.52 USD
77334-154 77342-256 77370-164 77382-234
Methyl 5-bromo-2-fluoro-3-(trifluoromethyl)benzoate ≥97%
Methyl 5-bromo-2-fluoro-3-(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₉H₅BrF₄O₂
MW: 301.0324 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34766093
CAS Number: 2092867-73-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR