4-Fluoro-3-formyl-5-(trifluoromethyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

75667-500MG 75667-1G 75667-5G 75667-25G
77334-152EA 202.91 USD
77334-152 77358-334 77370-162 77390-978
4-Fluoro-3-formyl-5-(trifluoromethyl)benzoic acid ≥97%
4-Fluoro-3-formyl-5-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₄F₄O₃
MW: 236.1198 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34766091
CAS Number: 2387319-56-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR