1-(2,3-Dichloropyridin-4-yl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

75605-500MG 75605-1G 75605-5G 75605-25G
77334-144EA 227.63 USD
77334-144 77358-326 77370-154 77390-970
1-(2,3-Dichloropyridin-4-yl)-2,2-difluoroethanone ≥95%
1-(2,3-Dichloropyridin-4-yl)-2,2-difluoroethanone

Precaución: for Research Use Only

Formula: C₇H₃Cl₂F₂NO
MW: 226.0076 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34766078
CAS Number: 2353311-06-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR